ENA514234408 Enamine

3-bromopyridine-2-sulfonyl fluoride

  • MDL number MFCD30180326
  • Cas Number : 1934834-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-248849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-248849
Availability details for
Quantity
  • Availability
  • Add to Cart