ENA514234448 Enamine

9-methylphenanthrene

  • MDL number MFCD00092279
  • Cas Number : 883-20-5
  • Molecular Weight : 192.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-253907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-253907
Availability details for
Quantity
  • Availability
  • Add to Cart