ENA514234455 Enamine

[(4-chlorophenyl)methyl][2-(diethylamino)ethyl]amine

  • MDL number MFCD00018751
  • Cas Number : 84434-83-3
  • Molecular Weight : 240.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-254381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-254381
Availability details for
Quantity
  • Availability
  • Add to Cart