ENA514234456 Enamine

[(4-chlorophenyl)methyl][3-(diethylamino)propyl]amine

  • MDL number MFCD03832191
  • Cas Number : 108010-87-3
  • Molecular Weight : 254.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-254383

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-254383
Availability details for
Quantity
  • Availability
  • Add to Cart