ENA514234469 Enamine

5-(chloromethyl)-3-cyclopropyl-1-methyl-1H-1,2,4-triazole

  • MDL number MFCD26938255
  • Cas Number : 145578-19-4
  • Molecular Weight : 171.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-255554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-255554
Availability details for
Quantity
  • Availability
  • Add to Cart