ENA514234472 Enamine

2-[3-(1,2,4-thiadiazol-5-yloxy)phenyl]acetic acid

  • MDL number MFCD21211335
  • Cas Number : 1478992-30-1
  • Molecular Weight : 236.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-255750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-255750
Availability details for
Quantity
  • Availability
  • Add to Cart