ENA514234501 Enamine

tert-butyl N-{3-hydroxyspiro[3.3]heptan-1-yl}carbamate

  • MDL number MFCD30180358
  • Cas Number : 1935054-64-0
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-258827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-258827
Availability details for
Quantity
  • Availability
  • Add to Cart