ENA514234507 Enamine

1-methyl-3-(oxan-4-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD30180364
  • Cas Number : 1619991-95-5
  • Molecular Weight : 210.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-258977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-258977
Availability details for
Quantity
  • Availability
  • Add to Cart