ENA514234517 Enamine

8-benzyl-3,8-diazabicyclo[3.2.1]octane-2,4-dione

  • MDL number MFCD17977250
  • Cas Number : 731771-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-259995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-259995
Availability details for
Quantity
  • Availability
  • Add to Cart