ENA514234530 Enamine

3-(piperidin-3-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD30180377
  • Cas Number : 1934622-87-3
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-261291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-261291
Availability details for
Quantity
  • Availability
  • Add to Cart