ENA514234539 Enamine

thieno[2,3-d]pyrimidine-4-carboxylic acid

  • MDL number MFCD30180383
  • Cas Number : 1955519-70-6
  • Molecular Weight : 180.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262180

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262180
Availability details for
Quantity
  • Availability
  • Add to Cart