ENA514234540 Enamine

2-(3-hydroxy-2-oxopropyl)-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD21876296
  • Cas Number : 35750-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141-142
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262183
Availability details for
Quantity
  • Availability
  • Add to Cart