ENA514234547 Enamine

6-chlorocinnolin-4-ol

  • MDL number MFCD21086122
  • Cas Number : 876-89-1
  • Molecular Weight : 180.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262482

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262482
Availability details for
Quantity
  • Availability
  • Add to Cart