ENA514234671 Enamine

(2-bromo-5-fluorophenyl)methanesulfonamide

  • MDL number MFCD12783427
  • Cas Number : 1182947-94-9
  • Molecular Weight : 268.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-272758

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-272758
Availability details for
Quantity
  • Availability
  • Add to Cart