ENA514234860 Enamine

4-fluoro-1-isocyanato-2-methylbenzene

  • MDL number MFCD18269641
  • Cas Number : 67191-92-8
  • Molecular Weight : 151.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-146815

Safety Information

signalword Danger
pcodes P260,P264,P270,P271,P280,P284,P301+P310,P304+P340,P305+P351+P338,P310,P320,P330,P337+P313,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-146815
pictograms GHS06
hstatements H301,H319,H330
Availability details for
Quantity
  • Availability
  • Add to Cart