ENA514235375 Enamine

3-(2-amino-1-hydroxyethyl)benzonitrile hydrochloride

  • MDL number MFCD30180728
  • Cas Number : 50916-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-297182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-297182
Availability details for
Quantity
  • Availability
  • Add to Cart