ENA514235579 Enamine

2-benzyl 1-tert-butyl 5-oxopyrrolidine-1,2-dicarboxylate

  • MDL number MFCD08277159
  • Cas Number : 942603-69-2
  • Molecular Weight : 319.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-296234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-296234
Availability details for
Quantity
  • Availability
  • Add to Cart