ENA514236123 Enamine

3-(trifluoromethyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD24391721
  • Cas Number : 1516757-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-297518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-297518
Availability details for
Quantity
  • Availability
  • Add to Cart