ENA514236634 Enamine

5-chloro-2-(oxane-3-carbonyl)aniline hydrochloride

  • MDL number MFCD30181155
  • Cas Number : 1955540-49-4
  • Molecular Weight : 276.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-301588

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-301588
Availability details for
Quantity
  • Availability
  • Add to Cart