ENA514236655 Enamine

1-(1,4-diazepan-1-yl)-3-methylbutan-1-one hydrochloride

  • MDL number MFCD30181182
  • Cas Number : 1955507-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-301981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-301981
Availability details for
Quantity
  • Availability
  • Add to Cart