ENA514236783 Enamine

6-(difluoromethyl)pyridine-3-carbonitrile

  • MDL number MFCD17170006
  • Cas Number : 206201-65-2
  • Molecular Weight : 154.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-300302

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-300302
Availability details for
Quantity
  • Availability
  • Add to Cart