ENA514236810 Enamine

3-methoxy-4-(5-methyl-1,3,4-oxadiazol-2-yl)aniline

  • MDL number MFCD21962134
  • Cas Number : 1239719-68-6
  • Molecular Weight : 205.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-300373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-300373
Availability details for
Quantity
  • Availability
  • Add to Cart