ENA514236814 Enamine

2-(pyrrolidin-2-yl)-4-(trifluoromethyl)pyrimidine hydrochloride

  • MDL number MFCD30181046
  • Cas Number : 1955557-37-5
  • Molecular Weight : 253.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-300397

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-300397
Availability details for
Quantity
  • Availability
  • Add to Cart