ENA514237010 Enamine

4-bromo-3-methylbenzene-1-carboximidamide

  • MDL number MFCD09746044
  • Cas Number : 202805-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-301140

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-301140
Availability details for
Quantity
  • Availability
  • Add to Cart