ENA514237223 Enamine

1-bromo-4-(difluoromethoxy)-2-(trifluoromethyl)benzene

  • MDL number MFCD28737937
  • Cas Number : 1805524-34-8
  • Molecular Weight : 291.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-302046

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-302046
Availability details for
Quantity
  • Availability
  • Add to Cart