ENA514237238 Enamine

5-fluoroisoquinoline-8-carboxylic acid

  • MDL number MFCD26667638
  • Cas Number : 1369177-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-298948

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-298948
Availability details for
Quantity
  • Availability
  • Add to Cart