ENA514237301 Enamine

(3-methoxy-1,2-thiazol-5-yl)methanesulfonamide

  • MDL number MFCD30181222
  • Cas Number : 1934806-57-1
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-302317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-302317
Availability details for
Quantity
  • Availability
  • Add to Cart