ENA514237312 Enamine

2-bromo-5-phenyl-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD24038553
  • Cas Number : 1019704-99-4
  • Molecular Weight : 284.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-302334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-302334
Availability details for
Quantity
  • Availability
  • Add to Cart