ENA514237350 Enamine

2-[3-(4-fluorophenyl)-6-oxo-1,6-dihydropyridazin-1-yl]acetic acid

  • MDL number MFCD07799238
  • Cas Number : 853318-09-9
  • Molecular Weight : 248.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-302375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-302375
Availability details for
Quantity
  • Availability
  • Add to Cart