ENA514237414 Enamine

2-(1-benzyl-2,5-dioxoimidazolidin-4-yl)acetic acid

  • MDL number MFCD28248844
  • Cas Number : 1790213-67-0
  • Molecular Weight : 248.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-302446

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-302446
Availability details for
Quantity
  • Availability
  • Add to Cart