ENA514237726 Enamine

methyl 2-(7-hydroxy-4-methyl-2-oxo-2H-chromen-3-yl)acetate

  • MDL number MFCD03661530
  • Cas Number : 95903-37-0
  • Molecular Weight : 248.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-302708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-302708
Availability details for
Quantity
  • Availability
  • Add to Cart