ENA514237891 Enamine

2-({7-methyl-4-oxo-1H,2H,3H,4H-cyclopenta[c]chromen-9-yl}oxy)propanoic acid

  • MDL number MFCD02058427
  • Cas Number : 314744-27-9
  • Molecular Weight : 288.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-302878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-302878
Availability details for
Quantity
  • Availability
  • Add to Cart