ENA514237908 Enamine

3-{3,5,9-trimethyl-7-oxo-7H-furo[3,2-g]chromen-6-yl}propanoic acid

  • MDL number MFCD03660307
  • Cas Number : 777857-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-302895

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-302895
Availability details for
Quantity
  • Availability
  • Add to Cart