ENA514237915 Enamine

2-{3,5-dimethyl-7-oxo-7H-furo[3,2-g]chromen-6-yl}acetic acid

  • MDL number MFCD03681290
  • Cas Number : 664366-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-302902

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-302902
Availability details for
Quantity
  • Availability
  • Add to Cart