ENA514237973 Enamine

2-[2-(3,4-dimethoxyphenyl)ethyl]-1,3-dioxo-2,3-dihydro-1H-isoindole-5-carboxylic acid

  • MDL number MFCD02905158
  • Cas Number : 356573-99-4
  • Molecular Weight : 355.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-302964

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-302964
Availability details for
Quantity
  • Availability
  • Add to Cart