ENA514238037 Enamine

[(2,3-dimethoxyphenyl)methyl][3-(dimethylamino)propyl]amine dihydrochloride

  • MDL number MFCD07109521
  • Cas Number : 1158577-04-8
  • Molecular Weight : 325.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-303031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-303031
Availability details for
Quantity
  • Availability
  • Add to Cart