ENA514238239 Enamine

4-[2-(trifluoromethoxy)phenyl]-3H,4H,5H,6H,7H-imidazo[4,5-c]pyridine

  • MDL number MFCD09996859
  • Cas Number : 1010878-91-7
  • Molecular Weight : 283.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-303236

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-303236
Availability details for
Quantity
  • Availability
  • Add to Cart