ENA514238305 Enamine

6,7-dihydro-5H-benzo[7]annulen-2-amine hydrochloride

  • MDL number MFCD30181255
  • Cas Number : 1955514-98-3
  • Molecular Weight : 195.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-303312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-303312
Availability details for
Quantity
  • Availability
  • Add to Cart