ENA514238393 Enamine

2-(3-chlorophenyl)-5,5-dimethylmorpholine

  • MDL number MFCD09901507
  • Cas Number : 1017396-25-6
  • Molecular Weight : 225.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-303642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-303642
Availability details for
Quantity
  • Availability
  • Add to Cart