ENA514506342 Enamine

tert-butyl 2,6-dimethylpiperidine-3-carboxylate

  • MDL number MFCD30342719
  • Cas Number : 1989659-68-8
  • Molecular Weight : 213.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-251302

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-251302
Availability details for
Quantity
  • Availability
  • Add to Cart