ENA514506347 Enamine

(6R,7aS)-6-hydroxy-2-methyl-hexahydro-1H-pyrrolo[1,2-c]imidazolidin-3-one

  • MDL number MFCD30342724
  • Cas Number : 1989638-21-2
  • Molecular Weight : 156.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-261858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-261858
Availability details for
Quantity
  • Availability
  • Add to Cart