ENA514506370 Enamine

tert-butyl 2-cyano-2-{[1,3]thiazolo[5,4-b]pyridin-2-yl}acetate

  • MDL number MFCD30342744
  • Cas Number : 1989672-32-3
  • Molecular Weight : 275.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-268309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268309
Availability details for
Quantity
  • Availability
  • Add to Cart