ENA514506427 Enamine

N1,N1-dimethylspiro[3.3]heptane-1,3-diamine dihydrochloride

  • MDL number MFCD30342786
  • Cas Number : 1989671-57-9
  • Molecular Weight : 227.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-304249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-304249
Availability details for
Quantity
  • Availability
  • Add to Cart