ENA514506437 Enamine

2-[(1H-1,2,3-triazol-1-yl)methyl]benzoic acid hydrochloride

  • MDL number MFCD30342794
  • Cas Number : 1989659-13-3
  • Molecular Weight : 239.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-304663

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-304663
Availability details for
Quantity
  • Availability
  • Add to Cart