ENA514506565 Enamine

methyl 3-(5-cyclopropyl-1,3,4-oxadiazol-2-yl)propanoate

  • MDL number MFCD30342895
  • Cas Number : 1989659-61-1
  • Molecular Weight : 196.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-306177

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-306177
Availability details for
Quantity
  • Availability
  • Add to Cart