ENA514506572 Enamine

2-(dichloromethyl)-5-methyl-1,3,4-thiadiazole

  • MDL number MFCD30342900
  • Cas Number : 75341-27-4
  • Molecular Weight : 183.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-306264

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-306264
Availability details for
Quantity
  • Availability
  • Add to Cart