ENA514506644 Enamine

2-methylidene-1lambda6-thiolane-1,1-dione

  • MDL number MFCD30342573
  • Cas Number : 79496-71-2
  • Molecular Weight : 132.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-299287

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-299287
Availability details for
Quantity
  • Availability
  • Add to Cart