ENA514506660 Enamine

[1-(ethylsulfanyl)cyclopropyl]methanamine

  • MDL number MFCD21127262
  • Cas Number : 1483740-60-8
  • Molecular Weight : 131.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-299943

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-299943
Availability details for
Quantity
  • Availability
  • Add to Cart