ENA514506690 Enamine

3,3-dimethyl-3,4-dihydro-2H-pyran-4-one

  • MDL number MFCD23703033
  • Cas Number : 625099-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-300988

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-300988
Availability details for
Quantity
  • Availability
  • Add to Cart