ENA514506697 Enamine

5-chloro-1-ethyl-1H-pyrazole

  • MDL number MFCD23134858
  • Cas Number : 1427020-01-6
  • Molecular Weight : 130.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-301318

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-301318
Availability details for
Quantity
  • Availability
  • Add to Cart